Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parter Medical Products™ Sterile Specimen Containers
Incorporate the proprietary PMP™ Sure-Seal™ integrated sealing system and the proprietary PMP Sure-Lok™ proof-of-closure system
Supplier: Parter Medical Products™ 242517
This item is not returnable.
View return policy
Description
- Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
- Stackable, leak-resistant and graduated unless otherwise noted
- Sterile containers labeled with a security tab label and sterilized by gamma irradiation to SAL 10-3; certification of sterility available upon request
- Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used is free from latex and bovine contamination
- Manufactured under strict process control and lot numbers are fully traceable
- 95kpa compliant; testing results available upon request
- Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
- Available in individually wrapped (Flo-Pac™) formats
- Closures secure at 10 to 12 in.-lb. closing force
- Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post-production cap-back-off
- Standard cap colors are natural, orange and blue
Specifications
Yes | |
Sterile | |
120 mL | |
Polyethylene | |
Tamper-evident Label |
Clarified Polypropylene | |
4 oz. | |
Blue | |
53 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title