Learn More
Newcomer Supply Paraffin Wax Trimmer - Digital XH-90D. Excellent for removing excess paraffin from embedding tissue cassettes. Can clean multiple tissue cassettes simuntaneously.
SDP
Supplier:  Newcomer Supply 6528
Encompass_Preferred
The digital paraffin wax trimmer XH-90D model from Premiere is an efficient appliance for removing excess paraffin from embedding tissue cassettes.  The digital temperature controls allows for the user to set the temperature of the heating plate from room temperature up to 120°C in one degree increments. You can trim multiple cassettes simultaneously on the grooved heating surface. Compact tabletop design of the digital paraffin trimmer fits easily in any lab. The melted paraffin is easily melted away and drains into the removable tray below. Collection tray stays in place by magnetic contact with unit. Also available is a convenient disposable tray liner that allows easy disposal of the runoff wax. No need to scrape out the collection tray! Overall size: 11.25" x 5" x 8". Plate size: 9" x 5".
Catalog No. NC0779509