Learn More
Cole-Parmer Dynalon Polypropylene Buchner funnel, 240 mm dia
Supplier: Cole-Parmer 0612820
Choose Buchner funnels for gravity or vacuum filtration. Recommended sterilization: HDPE for gas, PP by autoclave or gas.