Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxolinic Acid, EP, 98-102%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail

O2595, 14698-29-4, C13H11NO5

Spectrum™ Chemical Oxolinic Acid, EP
CAS 14698-29-4
CAS Max % 1
Loss on Drying 0.005
Molecular Formula C13H11NO5
MDL Number MFCD00056775
Packaging Poly Pail
Quantity 5 kg
InChI Key KYGZCKSPAKDVKC-UHFFFAOYSA-N
SMILES CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12
IUPAC Name 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid
Molecular Weight (g/mol) 261.23
Percent Purity 98 to 102%
Grade EP
Show More Show Less