Learn More
YSI 3P LifeScience OXITOP CALIBRATING TABLETS
Supplier: YSI 3P LifeScience 209333
System check of OxiTop systems with calibration tablets for checking 6 measuring units (long term test of leakage testing and negative pressure).