Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oxidation Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.05M Iodine in 80/10/10 Acetonitrile/Pyridine/Water (v/v)
$3972.00 - $13274.00
Specifications
CAS | 75-05-8, 7732-18-5, 110-86-1, 7553-56-2 |
---|---|
Boiling Point | 75.5°C (1.013 hPa) |
Flash Point | 8.9°C |
Packaging | Returnable Steel Drum |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
60-090-008
|
Honeywell Chemicals
BC76156 |
56 L |
Each for $3,972.00
|
|
|||||
60-090-006
|
Honeywell Chemicals
BC761200 |
200 L |
Each for $13,274.00
|
|
|||||
Description
Specifications
75-05-8, 7732-18-5, 110-86-1, 7553-56-2 | |
8.9°C | |
Liquid |
75.5°C (1.013 hPa) | |
Returnable Steel Drum | |
Oxidation reagents |
Safety and Handling
Recommended Storage : Room Temperature
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title