Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid TS, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle

O-098, 6153-56-6

Spectrum™ Chemical Oxalic Acid TS, (U.S.P. Test Solution) is a solution, sometimes designated with “TS or Test Solution”, that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS 6153-56-6, 7732-18-5
CAS Max % 0.001, 0.999
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES O.O.OC(=O)C(O)=O
IUPAC Name oxalic acid dihydrate
Molecular Weight (g/mol) 126.06
Grade Chemical Solution
Quantity 100 mL
Packaging Poly Bottle