Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ Spacer Sets for Aluminum-Backed Sequencers

Choose from two sizes for manual sequencing with Thermo Scientific™ Owl™ Spacer Sets for Aluminum-Backed Sequencers.

Supplier:  Thermo Scientific™ S2SLSA2

 View more versions of this product

Catalog No. OW-S2SL-SA2

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Choose from two sizes for manual sequencing with Thermo Scientific™ Owl™ Spacer Sets for Aluminum-Backed Sequencers.

Available in 1.0W x 45cmL (0.4mm thick) or 1.0W x 65cmL (0.2mm thick).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spacer Set; 1.0W x 65cmL; 0.2mm thick
65 cm
0.2 mm
Owl™ S3S Aluminum-Backed Sequencer System
Spacers
1.0 cm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit