Learn More
Thermo Scientific Overpressure Relief Valve 60BAR 1EA
Supplier: Thermo Scientific 6083.9260
Overpressure relief valve with VH-D1 valve configuration and 60 bar set pressure opens and vents excess fluid when system pressure exceeds 60 bar, protecting equipment from damage until pressure drops below valve's reseat pressure.