Learn More
VistaLab™ Technologies Ovation™ Adjustable-Volume Pipetters PROMO
Naturally follows the contours of the palm.
$441.00
Specifications
For Use With | Left- or Right Hand Users |
---|---|
Product Line | Ovation™ |
Display | LCD |
For Use With (Application) | Adjustable Hook Can be Easily Positioned for a Custom Fit for Left- or Right Hand Users |
Catalog Number | Mfr. No. | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Color | Price | Quantity & Availability | ||||
22-146-601TI
![]() |
VistaLab™ Technologies
10570010TI |
1 to 10 µL | Orange |
Each for $441.00
|
|
22-146-602TI
![]() |
VistaLab™ Technologies
10570020TI |
2 to 20 µL | Yellow |
Each for $441.00
|
|
22-146-603TI
![]() |
VistaLab™ Technologies
10570100TI |
10 to 100 µL | Green |
Each for $441.00
|
|
22-146-604TI
![]() |
VistaLab™ Technologies
10570200TI |
20 to 200 µL | Blue |
Each for $441.00
|
|
22-146-605TI
![]() |
VistaLab™ Technologies
10571000TI |
100 to 1000 µL | Blue |
Each for $441.00
|
|
Description
- The adjustable hook can be easily positioned for a custom fit for left- or right-handed users
- Fits comfortably in the palm of your hand
- Volume setting is indicated by an LCD readout on the pipetter handle
- Significantly improves postures and reduces forces associated with pipetting
- Five popular volume settings can be programmed and recalled by pressing a single button
- Energy release button stores the energy captured during the acquisition of tips and uses energy to push off the tip at the gentle push of a button
- Low-force, form-fitting plunger allows user to manually control the speed of aspirating and dispensing with minimal effort
- Nozzle with replaceable filter prevents contamination of the pipetter
- Stands securely on the counter before, during, and after the job—no risk of contamination
- In-lab calibration adjusted with internal software
Specifications
Left- or Right Hand Users | |
Ovation™ | |
LCD | |
Adjustable Hook Can be Easily Positioned for a Custom Fit for Left- or Right Hand Users |