Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VistaLab™ Technologies Ovation™ Adjustable-Volume Pipetters PROMO
Naturally follows the contours of the palm.
Supplier: VistaLab™ Technologies 10570100TI
This item is not returnable.
View return policy
Description
- The adjustable hook can be easily positioned for a custom fit for left- or right-handed users
- Fits comfortably in the palm of your hand
- Volume setting is indicated by an LCD readout on the pipetter handle
- Significantly improves postures and reduces forces associated with pipetting
- Five popular volume settings can be programmed and recalled by pressing a single button
- Energy release button stores the energy captured during the acquisition of tips and uses energy to push off the tip at the gentle push of a button
- Low-force, form-fitting plunger allows user to manually control the speed of aspirating and dispensing with minimal effort
- Nozzle with replaceable filter prevents contamination of the pipetter
- Stands securely on the counter before, during, and after the job—no risk of contamination
- In-lab calibration adjusted with internal software
Specifications
±0.8% at 100 μL, ±0.8% at 50 μL, ±3.5% (at 10 μL) | |
Left- or Right Hand Users | |
10 to 100 μL | |
Adjustable Hook Can be Easily Positioned for a Custom Fit for Left- or Right Hand Users |
Green | |
Ovation™ | |
LCD | |
0.1 μL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction