Learn More
PATTERSON DENTAL COMPANY Contemporary Ortho-Jet - Liquid, 8 oz Bottle
Supplier: PATTERSON DENTAL COMPANY 4594750
Contemporary Ortho from Jet Lang is a fast-curing orthodontic acrylic resin with the same properties as Ortho-Jet for the purpose of generating accurate orthodontic appliances, while adding a splash of color.