Learn More
Thermo Scientific™ Orion™ 8104BNUWP ROSS Ultra™ pH Electrode
Obtain precise pH determinations and a toughened bulb for rugged lab use with the Thermo Scientific™ Orion™ 8104BNUWP ROSS Ultra™ pH Electrode.
Supplier: Thermo Scientific™ 8104BNUWP
Description
Obtain precise pH determinations and a toughened bulb for rugged lab use with the Thermo Scientific™ Orion™ 8104BNUWP ROSS Ultra™ pH Electrode, a general purpose combination electrode with a top-performance glass body.
Includes:- One meter cable
Requires:
- pH buffers (pH 7 ±1 to 3 pH units) Beakers, plastic or glass
- Magnetic stirrer or Orion stirrer probe
- Distilled or deionized water
Compatible with:
BNWP connector is compatible with any standard BNC connector both on Thermo Scientific™ Orion Star™ Series meters and competitive meter models.
Recommended for:
- Rugged lab use and precise Q.C. measurements
Specifications
Glass combination electrode w/toughened bulb; 120 x 12mm | |
Ceramic | |
0 to 14 | |
pH Electrode | |
ROSS | |
32°F to 212°F | |
1 m |
WPBNC | |
Glass Body | |
0°C to 100°C | |
3M KCl | |
0.01 | |
39.4 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.