Learn More
Drummond™ Original Pipet-Aid™ Pipet Controller
Sets the standard for a safe and convenient alternative to mouth pipetting in the laboratory.
Supplier: Drummond™ 4000221TC
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
A patented, easy-to-use volume control mechanism permits precise fluid intake and delivery. The check valve protects against accidental fluid intake into the handle.
- Can be used with glass or plastic pipets
- Autoclavable components
- Optional EZ Grip Tissue Culture (TC) Nosepiece prevents cross-contamination due to over-pipetting
- Special dual pump filtration unit for use with biohazardous materials is available
- 220V CE models for use in Europe only (not for use in the U.S.A)
Order Info
Complete models include Pipet-Aid, TC nosepiece, four extra filters and dual-pump filtration unit. Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Specifications
EZ Grip TC Filter Nosepiece | |
Complete (Dual Pump, TC Nosepiece), CE/Eur | |
1 to 150 mL | |
Pipetting of biohazardous materials | |
220 V |
Autoclavable | |
Filler/Dispenser | |
Dual pump filter unit | |
Glass or plastic pipets |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.