Learn More
Thermo Scientific ORGANIC SPRAY CHAMBER
Supplier: Thermo Scientific 842312051311
Organics Spray Chamber; Spraychamber with baffle to reduce the amount of sample that reaches; the plasma.; Compatible with all iCAP 6000 Series Spectrometers.