Learn More
MilliporeSigma™ Supelco™ ORBO™ 33 Activated Petroleum Charcoal (20/40), 700/390 mg W,F,F separators
Used for gas and vapor sampling
Manufacturer:  MilliporeSigma™ Supelco™ 20259
Catalog No. 11-100-9142
Add to cart
Description
Description
ORBO™ 33 Activated Petroleum Charcoal (20/40), 700/390 mg solvent desorption tube is designed for gas and vapor sampling. It contains two beds of the same selective adsorbent separated by glass wool or foam. Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Specifications
Specifications
W,F,F Separators
50/Pk.
150 mm
8 mm
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
ORBO is a trademark of Sigma-Aldrich Co. LLC