Learn More
DNA Genotek™ ORAcollect•RNA
ORAcollect•RNA is a non-invasive, saliva self-collection and stabilization swab device for use in validated assays for the detection of the SARS-CoV-2 RNA virus. Used in laboratory-based tests for back-to-work and back-to-school programs, population screening, and laboratory testing.
Supplier: DNA Genotek™ ORE100
Alert:
For IVD, Rx, and For Use Under Emergency Use Authorization
Description
- Easy sampling method makes ORAcollect•RNA suitable for self or assisted saliva collection
- Inactivates the SARS-CoV-2 virus for safe sample transportation and handling
- Room temperature stability for human, bacterial, and viral RNA samples
- Compatible with many RNA extraction kits, e.g. Zymo Research, ThermoFisher Scientific, QIAGEN, etc.
- Integrates with automated laboratory equipment for high throughput sample processing
- Does not contain guanidinium for laboratory safety
- Validated for the detection and diagnosis of SARS-CoV-2 on multiple tests
Specifications
Saliva Collection Device | |
COVID-19 testing: Emergency Use Authorized | |
Non-sterile | |
1 mL |
FDA EUA Authorized, CE Marked, Health Canada Interim Order Authorization | |
Swab attached to double-ended cap, collection tube, and IFU | |
Room temperature |
Safety and Handling
ShelfLife : 12 months to 24 months