Learn More
McKesson General Medical Ora-Sweet SF 16 oz. Citrus Berry Flavor
Supplier: McKesson General Medical 867580
ORA-Sweet SF is a sugar-free, alcohol-free syrup vehicle used to simplify the process of flavoring and sweetening extemporaneous compounded sugar-free oral preparations. Will retain its flavoring properties when diluted up to 50 % with water or suspending agents.