Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPTIZYME™ SpeI (BcuI), Fisher BioReagents™
Description
5'...A^C T A G T...3'
3'...T G A T C^A...5'
Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 50 - 100%
- Buffer 2: 50 - 100%
- Buffer 3: 0 - 20%
- Buffer 4: 100%
- Buffer 5: 20 - 50%
- 10mM Tris-HCl (pH 7.5 at 25°C), 300mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
- A minimum of 10 units of SpeI is required for the complete digestion of 1μg of agarose-embedded Adenovirus-2 DNA in 16 hours.
Specifications
10 U/μL | |
37°C | |
>95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with SacI | |
400 U | |
SacI |
10X OPTIZYME™ Ecl136II, SacI Buffer | |
7.4 | |
10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol | |
A.CTAGT | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction