Learn More
Cole-Parmer Thermo Scientific 967901 ORP Standard, one 475 mL bottle
Supplier: Cole-Parmer 0573055
These stable, ready-to-use standards require no mixing and contain no organics or hazardous chemicals and can be used straight out of the bottle