Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ONO AE3 208
High affinity and selective EP4 antagonist
$334.50 - $1367.00
Specifications
Purity | >98% |
---|---|
Product Type | ONO AE3 208 |
Molecular Weight (g/mol) | 404.43 |
Inhibitors | ONO AE3 208 |
Description
High affinity and selective EP4 receptor antagonist (Ki values are 1.3, 30, 790 and 2400 nM for EP4, EP3, FP and TP receptors respectively). Displays no affinity for EP1, EP2, DP or IP receptors (Ki >10 μM). Inhibits PGE2-induced IL-8 production in colonic epithelial caco-2 cells and attenuates PGE2 inhibition of natural killer T cell activation. Suppresses recovery from experimentally-induced colitis and stimulates CD4+ T cell proliferation in C57BL/6 mice. Also reduces metastasis of mammary tumor cells in a murine model of breast cancer. Orally active.Specifications
>98% | |
ONO AE3 208 | |
404.43 | |
ONO AE3 208 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title