Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ One Click™ Melting Point System

One Click™ user interface minimizes training and operator influence. Mettler Toledo™ One Click™ Melting Point delivers a strong performance with the common user interface for all Analytical Laboratory instruments.

Manufacturer:  Mettler Toledo™ 51142502

Catalog No. 01-910-760

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Ideal choice for melting point and melting range determinations
  • Enhanced export possibilities
  • Color video recording
  • Large results storage
  • Customized user shortcuts
  • Quick and simple calibration
    • Automatic detection of transmission intensity
Specifications

Specifications

MP70 Melting Point System
Full compliance with the relevant international and national standards and norms, such as Ph.Eur., USP, Ch.P., and JIS is ensured
35 cm
7.4 in.
MP70
4 kg
18 cm
Ambient to 200°C: ±0.2°C;, 200°C to 350°C: ±0.5°C
13.7 in.
Fully automatic melting point (MP) and melting range (MR) determination. Pharmacopeia and Thermodynamic measuring modes
19 cm
10°C to 35°C
7.08 in.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit