Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Genotek™ OMNIgene™•ORAL OME-505 (For IVD, Rx and For Use Under Emergency Use Authorization)
A non-invasive, saliva self-collection and stabilization device for use in validated assays for the detection of the SARS-CoV-2 RNA virus (the coronavirus that causes COVID-19). DNA Genotek kits are used in laboratory-based tests for back-to-work and back-to-school programs, population screening applications, and laboratory testing.
Supplier: DNA Genotek™ OME505
Description
- FDA EUA authorized device for self or assisted collection of saliva for use in the detection of SARS-CoV-2 RNA
- Stabilization of SARS-CoV-2 RNA for at least 3 days at room temperature, eliminating the need for freezer storage and cold-chain transport
- Maintains sample integrity throughout temperature fluctuations encountered during real-life shipping and handling conditions (e.g., -20°C to +50°C)
- Safe for home collection with >99% inactivation of SARS-CoV-2 in collected and stabilized samples
- Improves donor compliance and maximizes successful sample collections with intuitive and validated instructions
- Bar coded for full sample traceability (128C format)
- Buffer solution does not include guanidine
- Compatible with commercially available extraction kits including Thermo Fisher, Zymo, and Omega Biotek
- Compatible with laboratory automation
Specifications
Saliva Collection Device | |
COVID-19 Testing: Emergency Use Authorized | |
8 Extractions/Kit, 200 Kits/Cs. | |
Room temperature |
FDA EUA Authorized, CE Marked, Health Canada Interim Order Authorization | |
Saliva funnel attached to collection tube, screw cap, packaging clamshell and IFU | |
Non-sterile | |
250 μL/Extraction |
Safety and Handling
ShelfLife : 24 months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title