Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OK-1™ Full Finger Anti-Vibration Gloves

Catalog Number p-92770
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves feature a pre-curved design for worn-in comfort from the first use. Anti-vibration technology protects hands by absorbing and dissipating vibrations caused by the constant use of power tools.

  • Premium grain leather with stretch nylon back
  • Designed to form to the natural curve of the hand for increased comfort
  • Machine washable
Color Brown
Hand Specific
Material Grain Cowhide Leather
Product Type Work Gloves
Size Large
For Use With (Application) Construction, Machine Operation, and Manufacturing
Glove Cuff Style Open
Series OK990
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit