Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Scout™ STX Portable Balances with Touchscreen Display
SCI_ED

The all new OHAUS™ Scout with intuitive color touchscreen interface offers superior overload protection in a sleek, low profile and stackable design.

Manufacturer:  OHAUS™ 30253016/EMD

 View more versions of this product

Catalog No. S14516


Add to cart

Description

Description

  • Fast response time and repeatable weighing results
  • Rapid stabilization and high resolution
  • Five major connectivity options
  • Additional software application modes and added weighing units complement the balance and set a new standard in portable weighing technology
  • Available with or without draftshield
  • Stackable design
  • Leveling bubble with adjustable feet
  • Kensington security slot cable sold separately
  • Calibration mass included(all models up to 620g)
  • 12 different weighing units:g,mg,kg,lb:oz,N,ct,Grn, dwt, ozt, custom unit
  • Powered by AC adapter (included) or 4 AA batteries (not included)
  • Integrated weigh-below hook
  • Transportation/storage lock
 
Applications
Weighing, density determination, molar mass
Specifications

Specifications

Color Touchscreen
8.2 kg
1 g
No draftshield
STX8200
1 sec.
Bluetooth, Ethernet, USB Host, USB Device, RS243
Touchscreen
Weighing, density determination, molar mass
ABS Housing, Stainless Steel Pan
Square
Portable Balance
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit