Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Octinoxate, USP, 95-105%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Octinoxate, USP absorbs ultraviolet radiation in the UVB range and is used in sunscreens and other cosmetics to protect from skin damage. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Acidity | To pass test |
CAS Max % | 1 |
Packaging | Amber Glass Bottle |
Refractive Index | 1.542-1.548 |
Quantity | 25 g |
Specific Gravity | 1.005-1.013 |
Percent Purity | 95 to 105% |
Grade | USP |