Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Octanoyl-L-carnitine-(N-methyl-d3), ≥97.0% (TLC), MilliporeSigma™ Supelco™
$429.00
Specifications
CAS | 204259-56-3 |
---|---|
Type | Analytical Standard |
Linear Formula | C15D3H26 NO4 |
MDL Number | MFCD01073498 |
Synonym | (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxooctyl)oxy]-1-propanaminium inner salt; L-Carnitine-(methyl-d3) octanoyl ester |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-3482
|
MilliporeSigma Supelco
5323010MG |
10 mg |
Each of 1 for $429.00
|
|
|||||
Description
Specifications
204259-56-3 | |
C15D3H26 NO4 | |
(2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxooctyl)oxy]-1-propanaminium inner salt; L-Carnitine-(methyl-d3) octanoyl ester | |
≥97.0% (TLC) |
Analytical Standard | |
MFCD01073498 | |
290.41 | |
Analytical Standard |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title