Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OccuNomix™ Embossed Rubber Back Anti-Vibration Gloves with VEP Padding
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Premium leather palm
- 4-way stretch spandex back
- Visco elastic polymer padding
- Hook & Loop wrist closure
- Terry cloth thumb to wipe away sweat
Specifications
Back Material | Spandex |
Color | Black |
Hand | Specific |
Material | Synthetic Leather |
Product Type | Anti-vibration Gloves |
Size | Small |
For Use With (Application) | Medium to Heavy Industrial Power Tools |
Glove Cuff Style | Hook and Loop |
Series | 426 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction