Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine, 0.1% Solution, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
CAS 612-82-8
Packaging Poly Bottle
Quantity 500 mL
Grade Chemical Solution