Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzyl-DL-serine, Thermo Scientific™

Click to view available options
Quantity:
5 g
5445-44-3
C10H13NO3
195.22
MFCD00021724,MFCD00065936
IDGQXGPQOGUGIX-UHFFFAOYNA-N
o-benzyl-dl-serine, 2-amino-3-benzyloxy propanoic acid, o-benzylserine, h-dl-ser bzl-oh, 3-benzyloxy-dl-alanine, benzylserine, 2-amino-3-phenylmethoxy propanoic acid, o-benzyl-serine, acmc-1artk, spectrum2_001351
4573
2-amino-3-phenylmethoxypropanoic acid
NC(COCC1=CC=CC=C1)C(O)=O
5445-44-3
C10H13NO3
195.22
MFCD00021724,MFCD00065936
IDGQXGPQOGUGIX-UHFFFAOYNA-N
o-benzyl-dl-serine, 2-amino-3-benzyloxy propanoic acid, o-benzylserine, h-dl-ser bzl-oh, 3-benzyloxy-dl-alanine, benzylserine, 2-amino-3-phenylmethoxy propanoic acid, o-benzyl-serine, acmc-1artk, spectrum2_001351
4573
2-amino-3-phenylmethoxypropanoic acid
NC(COCC1=CC=CC=C1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
O-Benzyl-DL-serine is used as catalytic agent, petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Store away from oxidizing agents.
Melting Point ∽225°C (decomposition)
Beilstein 3204905
Solubility Information Insoluble in water.
Formula Weight 195.22
Quantity 5 g
Chemical Name or Material O-Benzyl-DL-serine

EINECSNumber : 226-650-7

TSCA : No

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit