Learn More
Polysciences, Inc. Nylon Wool Fiber 50 g Used to separate leukocytes from peripheral blood and to separate T-cells from B-cells Handling: Gloves Storage: Store at room temperature
Supplier: Polysciences, Inc. 1836950
Nylon Wool Fiber 50 g Used to separate leukocytes from peripheral blood and to separate T-cells from B-cells Handling: Gloves Storage: Store at room temperature