Learn More
Lonza Walkersville MycoZap Mycoplasma Elimination Reagent (5 Treatments)
Supplier: Lonza Walkersville LT07918
Where contamination has occurred, and the sample cannot be discarded for certain reasons, the MycoZap Mycoplasma Elimination Reagent can clear detectable mycoplasma contamination in as few as 4 days. The MycoZap Reagent has been optimized to eliminate mycoplasma with minimal toxic effects on the infected cells. It eliminates mycoplasma by using a combination of antibiotic and antimetabolic agents. This approach allows for a highly reliable elimination of mycoplasma that cannot be achieved by the use of antibiotics alone. The MycoZap Reagent can be used to eradicate mollicutes, including Mycoplasma, Acholeplasma, Spiroplasma and Entomoplasma species in cell cultures.Benefits:Efficient mollicute elimination by a combination of antibiotic and antimetabolic agentsMinimal toxic effectsApplications:Eradicates Mycoplasma, Acholeplasma, Spiroplasma, and EntomoplasmaSuited for a broad range of cell culturesContent:1 x 2.5 mL MycoZap Reagent 11 x 7.5 mL MycoZap Reagent 2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.