Learn More
MilliporeSigma NUCLEAR EXTRACTION KIT
Supplier: MilliporeSigma 2900
Nuclear Extraction Kit, Contains Cytoplasmic Lysis buffer, Nuclear extraction buffer, PBS packets, DTT, 1M, Protease inhibitor cocktail, Detergent, 10%, Nuclear extraction kit is shipped and stored at -20 deg.C, 100 extractions