Learn More
MilliporeSigma™ Novagen™ Origami™ 2(DE3) Singles™ Competent Cells
Origami 2 strains have mutations in glutathione reducatase (gor) and thioredoxin reductase (trxB), facilitating proper disulfide bond formation.
$253.00 - $441.00
Specifications
Efficiency Guaranteed efficiency: 2 x 106 cfu/μg
Content And Storage ≤-70°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
71-408-4
View Documents
MilliporeSigma™
714084
22 Reactions
Each for $441.00
Add to Cart
71-408-3
View Documents
MilliporeSigma™
714083
11 Reactions
Each for $253.00
Add to Cart
Description
Description
Origami™ 2 host strains are K-12 derivatives that have mutations in both the thioredoxin reductase (trxB) and glutathione reductase (gor) genes, which greatly enhance disulfide bond formation in the E. coli cytoplasm. The Origami 2 strains are kanamycin sensitive; like the original Origami strains, the gor mutation is still selected for by tetracycline. To reduce the possibility of disulfide bond formation between molecules, strains containing mutations in trxB and gor are recommended only for the expression of proteins that require disulfide bond formation for proper folding. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Specifications
Specifications
Guaranteed efficiency: 2 x 106 cfu/μg
≤-70°C