Learn More
MilliporeSigma™ Novagen™ Mouse Genomic DNA
For genomic analysis, including PCR and library construction. It is free of contaminating RNA and protein.
Supplier: MilliporeSigma™ 692393/DEL
This item is not returnable.
View return policy
Description
- High molecular weight
- High purity DNA for any application
- Isolation protocol minimizes shearing and removes contaminants that inhibit enzymatic procedures
- Pipet with a wide-bore pipet tip to minimize shearing
- At least 90% of DNA is greater than 100kbp in size, as analyzed by CHEF gel electrophoresis (>50kbp for S. cerevisiae)
- No inhibition of restriction enzyme activity
Specifications
8 | |
10mM Tris-HCl, 1mM EDTA (TE) | |
2°C to 8°C | |
PCR, Library construction |
Mouse Genomic DNA | |
Multiple organisms | |
100 μg |