Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Novagen™ EMSA Accessory Kit
Used for testing DNA binding proteins.
Supplier: MilliporeSigma™ 712823
Description
EMD Millipore Novagen™ EMSA Accessory Kit determines whether specific DNA sequence binds to protein in electrophoretic mobility shift assays.
- Contains four key reagents
- Compatible with traditional methods of nuclear extract isolation
- Provides sufficient reagents to perform 100 reactions
Specifications
Store at -20°C. | |
Poly(dI-dC) Poly(dI-dC) Solution, 125 uL; 4X Binding Buffer, 1 mL; Sonicated Salmon Sperm DNA, 150 uL; 100 mM DTT, 100 uL | |
Plastic ampoule |
NucBuster™ Protein Extraction Kit | |
100 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction