Learn More
Honeywell™ North™ Survivair™ Premier™ Plus Half-Mask Respirator, S-Series, Medium/Large
Silicone half-mask respirator is designed with a triple flange design for superior fit, comfort and protection
Supplier: Honeywell™ 322500
This item is not returnable.
View return policy
Description
Survivair Premier Plus facepiece assembly.
- S-Series; Size: Medium/Large
- Three flanges; provides “backup” sealing security
- Silicone facepiece construction is flexible, lightweight and durable
- Wide, wraparound and pleated silicone facepiece fits a large variety of facial types
- Strategic lateral placement of inhalation valves and cartridge ports enhances field of vision
- Exhalation valve provides quick exhaust of warm exhaled air
- Cradle suspension with D-ring roller mechanism for easy and secure adjustment
- Packaged in a resealable bag to protect the respirator from contamination when not in use
- OV/N95 (S-Series) cartridges/filters available with respirator for quick ordering
- Hazards: Airborne Particulates, Biohazard, Chemical, Contamination, Extreme Environment, Gas, Vapors, Smoke
Applications:
Chemical, Construction, Fire Protection, Government, Law Enforcement, Manufacturing, Medical, Military, Oil and Gas, Pharmaceutical, Utilities.
Compliance
NIOSH 42 CFR 84.
Warranty and Services
Free of defects in materials and workmanship for five years (one year for plastic parts) from the date of purchase.
Specifications
NIOSH 42 CFR Part 84 | |
Half-Mask Respirator | |
S Series Cartridge |
Silicone | |
Medium/Large | |
322500 |