Learn More
Honeywell™ North™ Premier 611 Half-Mask Respirators for Paints and Pesticides
Silicone half-mask respirators can be used with N95 or P95 cartridges and filters depending on specific environment.
Specifications
Material | Silicone |
---|---|
Product Type | Half-Mask Respirator |
For Use With (Application) | Chemical, construction, food services, forestry, government, manufacturing, military, mining, municipal services, oil and gas, pharmaceutical, steel and metals, welding |
For Use With (Equipment) | T Series Cartridge |
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity & Availability | ||||
17-631-5
|
Honeywell™
312120 |
Medium | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
17-631-3
|
Honeywell™
313110 |
Large | Cartridges and Filter | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||
17-631-6
|
Honeywell™
313120 |
Large | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Complies with NIOSH 42 CFR 84 standards
- Half-mask design same as Series 6100 respirator
- Easy-to-clean silicone
- Dual-cartridge features a soft sealing flange and pliable nosepiece for maximum comfort
- Nonslip cradle suspension and floating yoke ensure a secure fit
- Additional room for protective eyewear
- P95 filter for paints and pesticides containing oil; the N95 filter is suitable for non-oil environments
Specifications
Silicone | |
Half-Mask Respirator | |
Chemical, construction, food services, forestry, government, manufacturing, military, mining, municipal services, oil and gas, pharmaceutical, steel and metals, welding | |
T Series Cartridge |