Learn More
Jackson Immuno Research Labs Normal Sheep Serum, Unconjugated, 10ml
Supplier: Jackson Immuno Research Labs 013000121
Normal Sheep Serum, 10mL, Physical State: Freeze-dried powder, Protein Concentration: 60mg/mL, Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH: 7.6, Store product at 2 to 8degreeC