Learn More
Tocris Bioscience™ Nociceptin
Endogenous NOP agonist
Supplier: Tocris Bioscience™ 0910/1
This item is not returnable.
View return policy
Description
Nociceptin is an endogenous ligand for the NOP opioid receptor (pKi = 8.6). Has no significant activity at μ, δ or κ opioid receptors (pKi < 6).Specifications
Nociceptin | |
Nociceptin | |
Phe-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-Leu-Ala-Asn-Gln |
1809 | |
1 mg |