Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nochromix™ Reagent

Inorganic oxidizer chemically cleans glassware.
Supplier: Alconox 2450
Description
- Contains no metal ions
- Rinses freely leaving no metal residue
- Valuable for trace analysis, enzymology and tissue culture work
- Each hermetically sealed 3 oz. packet should be mixed with 1 gal. (3.7L) of concentrated sulfuric acid
Specifications
Crystalline | |
Hermetically sealed packet | |
3 oz. |
White | |
Chemically cleans Glassware for trace analysis, enzymology and Tissue Culture work |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title