Learn More
New England Biolabs, Inc. NiCo21(DE3) Competent E. coli – 20 x 0.05 ml/tube
Supplier: New England Biolabs, Inc. C2529H
Chemically competent E. coli cells suitable for T7 expression with improved purity of target proteins isolated by IMAC.