Learn More
LECO Corporation Nickel Boat Liners, 100 per pack
Supplier:  LECO Corporation 502343
Encompass
Nickel Boat Liner - For use with 529-203 and 528-203-250 Ceramic Boats of TruMac Series. Size (MM): 50 x 15 x 8. 100/pk
Catalog No. NC9538390