Learn More
Fine Science Tools Moria Nickel Plated Pin Holder, 1EA
Supplier: Fine Science Tools 2601612
This pin holder works well with all pins and tungsten needles. It has a jaw opeing diameter of 0-1mm. The total length of this item is 12cm.