Learn More
Cliawaived Inc NicCheck Cotinine Control Set (2 x 12mL- 1 Postive/ 1 Negative)
Supplier: Cliawaived Inc NICCHECK CONTROLS
NicCheck Positive/Negative Cotinine control set in human urine. For use with Nicheck I Test Strips. Contains 0.1% Thimerosol. 1-Positive 15 ug/mL (Part# 600-202), 1-Negative 0 ug/mL. Controls can be shipped at room temperature. Controls can be left out during the day if need be (performing screening tests) but should be returned to the refrigerator when not in use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.