Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nextteq™ Gastec™ Detector Tubes: Amines
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
$160.30 - $2231.00
Specifications
Components Measured | Amines |
---|---|
No. of Strokes | 1 |
Quantity | 10 |
Response Time | 1 min. |
Temperature Range (English) | 32°F to 104°F |
Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | No. of Strokes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | No. of Strokes | Price | Quantity & Availability | ||
19-026-219
|
Nextteq™
180 |
Amines | 5 to 100 | 1 min. | 1 |
Pack of 10 for $160.30
Case of 10 PK for $1,299.00
|
|
||
19-026-220
|
Nextteq™
180L |
Amines | 0.5 to 10 | 1 min. | 1 |
Case of 100 for $2,231.00
|
|
||
Description
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.Specifications
Amines | |
10 | |
32°F to 104°F |
1 | |
1 min. | |
0°C to 40°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title