Learn More
Labconco™ Neutrodine™ Molecular Filter
SDPSCI_ED
For use with Protector™ Echo™ and Airo™ filtered fume hood. Labconco Neutrodine™ Molecular Filter for multi-purpose use of acids/bases/solvents, separated or combined.
Supplier:  Labconco™ 9577400/EMD
Catalog No. S28323
This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
Description
Description
  • Molecular carbon filter unit for multi-purpose use
  • Has been tested and found efficient on over 500 chemicals
  • Comprehensive molecular filter that allows the simultaneous handling of solvents, acids, bases, ammonia and formaldehyde
  • May last two years or more before requiring replacement
  • Allows Protector Filtered Fume Hoods to act as fully-featured, containment-enhancing fume hoods without any ducting
Warranty and Services
One year
Specifications
Specifications
Carbon
4 in.
18 Kg
30 in.
41 x 76 x 10 cm (16 x 30 x 4 in.)
76 cm
Multi-purpose: Acids, Bases, Solvents, Acid-Sulfur, Ammonia-Amine, Formaldehyde-Formalin, Organic Vapor, Trace Odor
Protector™ Echo™ and Airo™ Filtered Fume Hoods
40 lb.
16 in.
10 cm
41 cm
Neutrodine Molecular Filter
Product Suggestions
Product Suggestions