Learn More
Eisco™ NeuLog™ Sound Kit
The Eisco™ NeuLog™ Sound Kit contains equipment to produce sound and measure it through a sound sensor. It is difficult for students to grasp the relation between generated sound and wave’s propagation. With this kit, students measure sound waves generated by tuning forks and a recorder and also isolate sound using different materials
Supplier: Eisco™ SNDKIT
Description
- Study sound isolation through various materials
- Observe 'beat' sounds
- Study sound level
- Study sound waves
- NeuLog Sound Logger Sensor NOT included
Specifications
426 Hz tuning fork; 480 Hz tuning fork; 512 Hz tuning fork; Two resonance boxes (wooden boxes); Hammer; Recorder (musical instrument); Sound isolation sponge; Felt; Ten paper towels; Sound isolation box; Egg tray; Sellotape | |
For use with sound logger sensor and computer. |