Learn More
Eisco™ NeuLog Light Kit
Allows students to investigate the electricity generated from the irradiation of light. EISCO NeuLog Light Kit is used to understand the concepts of conversion of sunlight energy directly into electrical energy.
Supplier: Eisco™ LGTKIT
Requires : Light logger sensor and voltage logger sensor
Description
Experiment kit for understanding the concepts to electrical energySpecifications
NeuLog Light Kit | |
Photovoltaic cell, two microscope slides, utility stand, right angle clamp, extension clamp, ruler, sellotape, four colored flashlights (colorless, blue, green, and red) with batteries, four colored filters (colorless, blue, green, and red), white paper, black paper, grey paper, and thin box for shadow experiments |