Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nessler's Reagent, Solution, For Nitrogen (Ammonia), APHA 18, Spectrum™ Chemical
SDP

Catalog Number 18604772
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1310-73-2, 7774-29-0, 7681-11-0, 7332-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]
1310-73-2, 7774-29-0, 7681-11-0, 7332-18-5
HNaO
40.00
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
[OH-].[Na+]

N-120

Spectrum™ Chemical Nessler's Reagent, Solution, For Nitrogen (Ammonia), APHA 18 - As an APHA certified solution, this item meets the ASTM D1209 standards for use in water and waste water treatment facilities to determine the concentration of dissolved and particulate materials. Spectrum Chemical manufactured APHA grade products meet the toughest regulatory standards for quality and purity.

CAS Max % 0.16, 0.1, 0.07, 0.67
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit